1H,4H,5H,6H-Cyclopenta[c]pyrazole-3-carboxamide ≥97%

Supplier: Matrix Scientific

076624-500MG 076624-1G 076624-5G
102547-152EA 803.08 USD
102547-152 102547-154 102547-156
1H,4H,5H,6H-Cyclopenta[c]pyrazole-3-carboxamide ≥97%
1H,4H,5H,6H-Cyclopenta[c]pyrazole-3-carboxamide

Hazard: Irritant

Formula: C₇H₉N₃O
Melting Pt: 215…217 °C
MDL Number: MFCD11506254
CAS Number: 851776-30-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR