5-(4-Chlorophenyl)pyridin-2-amine ≥97%

Supplier: Matrix Scientific

076622-500MG 076622-1G
102547-144EA 258.46 USD
102547-144 102547-146
5-(4-Chlorophenyl)pyridin-2-amine ≥97%
5-(4-Chlorophenyl)pyridin-2-amine

Hazard: Irritant

Formula: C₁₁H₉ClN₂
Melting Pt: 132…134 °C
MDL Number: MFCD06802135
CAS Number: 84596-08-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR