3-Chloro-5-(ethoxycarbonyl)pyridine-2-carboxylic acid ≥97%

Supplier: Matrix Scientific

076618-500MG 076618-1G
102547-126EA 1243.08 USD
102547-126 102547-128
3-Chloro-5-(ethoxycarbonyl)pyridine-2-carboxylic acid ≥97%
3-Chloro-5-(ethoxycarbonyl)pyridine-2-carboxylic acid

Hazard: Irritant

Formula: C₉H₈ClNO₄
Melting Pt: 125…127 °C
MDL Number: MFCD22373655
CAS Number: 1200497-77-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR