1-(3-Bromophenyl)-4,5,6,7-tetrahydro-1H-indazol-4-one ≥97%

Supplier: Matrix Scientific

076617-500MG 076617-1G 076617-5G
102547-120EA 298.46 USD
102547-120 102547-122 102547-124
1-(3-Bromophenyl)-4,5,6,7-tetrahydro-1H-indazol-4-one ≥97%
1-(3-Bromophenyl)-4,5,6,7-tetrahydro-1H-indazol-4-one

Hazard: Irritant

Formula: C₁₃H₁₁BrN₂O
Melting Pt: 115…118 °C
MDL Number: MFCD15524938
CAS Number: 1260832-62-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR