5-Amino-3,4-dihydro-2H-1,4-benzoxazin-3-one ≥97%

Supplier: Matrix Scientific

Synonyms: 5-Amino-2H-1,4-benzoxazin-3(4H)-one

076610-500MG 076610-1G 076610-5G
102547-082EA 276.92 USD
102547-082 102547-084 102547-086
5-Amino-3,4-dihydro-2H-1,4-benzoxazin-3-one ≥97%
5-Amino-3,4-dihydro-2H-1,4-benzoxazin-3-one

Hazard: Irritant

Formula: C₈H₈N₂O₂
MW: 164.17 g/mol
Melting Pt: 234…237 °C
MDL Number: MFCD11100025
CAS Number: 148890-63-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR