4-Methylpiperazin-2-one ≥95%

Supplier: Matrix Scientific

Synonyms: 1-Methyl-3-oxopiperazine

076606-1G 076606-5G 076606-25G
102547-070EA 107.69 USD
102547-070 102547-072 102552-040
4-Methylpiperazin-2-one ≥95%
4-Methylpiperazin-2-one

Hazard: Irritant/Moisture Sensitive

Formula: C₅H₁₀N₂O
MW: 114.15 g/mol
Melting Pt: 84…90 °C
MDL Number: MFCD11870124
CAS Number: 34770-60-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR