Methyl-1-aminocyclobutanecarboxylate hydrochloride ≥95.0%

Supplier: Matrix Scientific

076605-250MG 076605-1G 076605-5G
102547-064EA 427.69 USD
102547-064 102547-066 102547-068
Methyl-1-aminocyclobutanecarboxylate hydrochloride ≥95.0%
Methyl-1-aminocyclobutanecarboxylate hydrochloride

Hazard: Irritant

Formula: C₆H₁₂ClNO₂
MDL Number: MFCD04115288
CAS Number: 92398-47-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR