1,4-Diiodo-2,5-dimethoxybenzene ≥95.0%

Supplier: Matrix Scientific

076604-1G 076604-5G 076604-10G
102547-058EA 284.62 USD
102547-058 102547-060 102547-062
1,4-Diiodo-2,5-dimethoxybenzene ≥95.0%
1,4-Diiodo-2,5-dimethoxybenzene

Hazard: Irritant

Formula: C₈H₈I₂O₂
MW: 389.9569 g/mol
MDL Number: MFCD00461369
CAS Number: 51560-21-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR