Methyl-1-aminocyclopentanecarboxylate hydrochloride ≥95.0%

Supplier: Matrix Scientific

Synonyms: 1-Aminocyclopentanecarboxylic Acid Methyl Ester Hydrochloride, Methyl 1-aminocyclopentanecarboxylate hydrochloride, Methyl 1-Aminocyclopentanecarboxylate HCl, Cycloleucine Methyl Ester Hydrochloride

076601-1G 076601-5G 076601-25G
102547-042EA 89.23 USD
102547-042 102547-044 102664-104
Methyl-1-aminocyclopentanecarboxylate hydrochloride ≥95.0%
Methyl-1-aminocyclopentanecarboxylate hydrochloride

Hazard: Irritant

Formula: C₇H₁₃NO₂
MDL Number: MFCD03840364
CAS Number: 60421-23-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR