Methyl quinoxaline-5-carboxylate ≥95%

Supplier: Matrix Scientific

076599-250MG 076599-1G 076599-5G
102547-032EA 355.38 USD
102547-032 102547-034 102547-036
Methyl quinoxaline-5-carboxylate ≥95%
Methyl quinoxaline-5-carboxylate

Hazard: Irritant

Formula: C₁₀H₈N₂O₂
MDL Number: MFCD19703859
CAS Number: 6924-71-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR