(2-Methoxyphenyl)methanamine hydrochloride ≥95%

Supplier: Matrix Scientific

076598-10G 076598-100G
102547-028EA 38.46 USD
102547-028 102547-030
(2-Methoxyphenyl)methanamine hydrochloride ≥95%
(2-Methoxyphenyl)methanamine hydrochloride

Hazard: Irritant

Formula: C₈H₁₂ClNO
MDL Number: MFCD22192496
CAS Number: 42365-52-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR