Pyrroloquinoline quinone ≥95%

Supplier: Matrix Scientific

Synonyms: 4,5-Dioxo-4,5-dihydro-1H-pyrrolo[2,3-f]quinoline-2,7,9-tricarboxylic acid, 4,5-Dihydro-4,5-dioxo-1H-pyrrolo[2,3-f]quinoline-2,7,9-tricarboxylic acid, PQQ, Methoxatin

076597-250MG 076597-1G
102547-024EA 284.62 USD
102547-024 102547-026
Pyrroloquinoline quinone ≥95%
Pyrroloquinoline quinone

Hazard: Irritant/Refrigerate

Formula: C₁₄H₆N₂O₈
MW: 330.21 g/mol
MDL Number: MFCD00043125
CAS Number: 72909-34-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR