4-Chloro-3-(trifluoromethoxy)benzyl bromide ≥95%

Supplier: Matrix Scientific

Synonyms: 4-(Bromomethyl)-1-chloro-2-(trifluoromethoxy)benzene

076596-1G 076596-5G 076596-10G
102547-018EA 273.85 USD
102547-018 102547-020 102547-022
4-Chloro-3-(trifluoromethoxy)benzyl bromide ≥95%
4-Chloro-3-(trifluoromethoxy)benzyl bromide

Hazard: Irritant

Formula: C₈H₅BrClF₃O
MW: 289.48 g/mol
MDL Number: MFCD06660249
CAS Number: 886500-93-2
UN: 3265
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR