7-Nitroisatin ≥95%

Supplier: Matrix Scientific

Synonyms: 7-Nitroindoline-2,3-dione

076595-1G 076595-5G
102547-012EA 167.69 USD
102547-012 102547-014
7-Nitroisatin ≥95%
7-Nitroisatin

Hazard: Irritant

Formula: C₈H₄N₂O₄
MW: 192.1300 g/mol
Density: 1.609
MDL Number: MFCD02066502
CAS Number: 112656-95-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR