2-Amino-4,5-dichlorobenzoic acid ≥95%

Supplier: Matrix Scientific

076594-1G 076594-5G 076594-10G
102547-006EA 640 USD
102547-006 102547-008 102547-010
2-Amino-4,5-dichlorobenzoic acid ≥95%
2-Amino-4,5-dichlorobenzoic acid

Hazard: Irritant

Formula: C₇H₅Cl₂NO₂
MDL Number: MFCD08457795
CAS Number: 20776-61-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR