5-Methylindolin-2-one ≥95%

Supplier: Matrix Scientific

Synonyms: 5-Methyl-2-oxindole

076593-250MG 076593-1G
102547-002EA 924.62 USD
102547-002 102547-004
5-Methylindolin-2-one ≥95%
5-Methylindolin-2-one

Hazard: Irritant

Formula: C₉H₉NO
MW: 147.18 g/mol
MDL Number: MFCD02379906
CAS Number: 3484-35-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR