2,2,5,5-Tetramethylpiperazine dihydrochloride ≥95%

Supplier: Matrix Scientific

076592-250MG 076592-1G
102546-998EA 1209.23 USD
102546-998 102547-000
2,2,5,5-Tetramethylpiperazine dihydrochloride ≥95%
2,2,5,5-Tetramethylpiperazine dihydrochloride

Hazard: Irritant

Formula: C₈H₂₀Cl₂N₂
MW: 215.17 g/mol
MDL Number: MFCD22194320
CAS Number: 858851-27-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR