1,2,3,4-Tetrahydro-2,7-naphthyridine hydrochloride ≥95%

Supplier: Matrix Scientific

076590-250MG 076590-1G
102546-988EA 1395.38 USD
102546-988 102546-990
1,2,3,4-Tetrahydro-2,7-naphthyridine hydrochloride ≥95%
1,2,3,4-Tetrahydro-2,7-naphthyridine hydrochloride

Hazard: Irritant

Formula: C₈H₁₁ClN₂
MDL Number: MFCD18253770
CAS Number: 1354940-72-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR