5-Chlorosalicylic acid ≥95%

Supplier: Matrix Scientific

Synonyms: CSA, 5-Chloro-2-hydroxybenzoic acid

076588-10G 076588-100G
102546-980EA 35.38 USD
102546-980 102546-982
5-Chlorosalicylic acid ≥95%
5-Chlorosalicylic acid

Hazard: Irritant

Formula: C₇H₅ClO₃
MW: 172.57 g/mol
MDL Number: MFCD00002457
CAS Number: 321-14-2
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR