3,5-Dichloro-4-hydroxybenzaldehyde ≥95%

Supplier: Matrix Scientific

076585-1G 076585-5G
102546-966EA 73.85 USD
102546-966 102546-968
3,5-Dichloro-4-hydroxybenzaldehyde ≥95%
3,5-Dichloro-4-hydroxybenzaldehyde

Hazard: Irritant

Formula: C₇H₄Cl₂O₂
MW: 191.01 g/mol
Melting Pt: 156…158 °C
MDL Number: MFCD00017605
CAS Number: 2314-36-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR