6-Bromo-3-hydroxypicolinic acid ≥95%

Supplier: Matrix Scientific

076584-1G 076584-5G
102546-962EA 506.15 USD
102546-962 102546-964
6-Bromo-3-hydroxypicolinic acid ≥95%
6-Bromo-3-hydroxypicolinic acid

Hazard: Irritant

Formula: C₆H₄BrNO₃
MDL Number: MFCD18072573
CAS Number: 321596-58-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR