Dipyridamole ≥95%

Supplier: Matrix Scientific

Synonyms: 2,6-Bis(diethanolamine)-4,8-dipiperidinopyrimido[5,4-d]pyrimidine, 2,2',2'',2'''-((4,8-Di(piperidin-1-yl)pyrimido[5,4-d]pyrimidine-2,6-diyl)bis(azanetriyl))tetrakis(ethan-1-ol), 2,2',2'',2'''-[(4,8-Dipiperidinylpyrimido[5,4-d]pyrimidine-2,6-diyl)dinitrilo]tetrakisethanol

076583-5G 076583-10G 076583-100G
102546-954EA 66.15 USD
102546-954 102546-956 102546-958
Dipyridamole ≥95%
Dipyridamole

Hazard: Irritant

Formula: C₂₄H₄₀N₈O₄
MW: 504.64 g/mol
MDL Number: MFCD00010555
CAS Number: 58-32-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR