(S)-tert-Butyl-2-(aminomethyl)morpholine-4-carboxylate oxalate ≥95%
Supplier: Matrix Scientific
076582-250MG
076582-1G
102546-950EA
1209.23
USD
102546-950
102546-952
(S)-tert-Butyl-2-(aminomethyl)morpholine-4-carboxylate oxalate ≥95%
(S)-tert-Butyl-2-(aminomethyl)morpholine-4-carboxylate oxalate
Hazard: Irritant
Formula:
C₁₂H₂₂N₂O₇ MW: 306.32 g/mol |
MDL Number:
MFCD22194319 CAS Number: 1373255-08-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...