Methyl-5-bromoquinoline-2-carboxylate ≥95%

Supplier: Matrix Scientific

Synonyms: Methyl 5-bromoquinoline-2-carboxylate

076580-250MG 076580-1G
102546-940EA 924.62 USD
102546-940 102546-942
Methyl-5-bromoquinoline-2-carboxylate ≥95%
Methyl 5-bromoquinoline-2-carboxylate

Hazard: Irritant

Formula: C₁₁H₈BrNO₂
MW: 266.09 g/mol
MDL Number: MFCD21086458
CAS Number: 1355174-78-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR