3-Bromophthalic anhydride ≥95%
Supplier: Matrix Scientific
Synonyms:
4-Bromoisobenzofuran-1,3-dione
076579-5G
076579-25G
102546-936EA
238.46
USD
102546-936
102833-928
3-Bromophthalic anhydride ≥95%
3-Bromophthalic anhydride
Hazard: Irritant
Formula:
C₈H₃BrO₃ Melting Pt: 132-134°C Density: 1.911 |
MDL Number:
MFCD09800402 CAS Number: 82-73-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...