6-Fluoro-3-iodo-1H-pyrrolo[2,3-b]pyridine ≥95%

Supplier: Matrix Scientific

076578-250MG 076578-1G
102546-930EA 1395.38 USD
102546-930 102546-932
6-Fluoro-3-iodo-1H-pyrrolo[2,3-b]pyridine ≥95%
6-Fluoro-3-iodo-1H-pyrrolo[2,3-b]pyridine

Hazard: Irritant

Formula: C₇H₄FIN₂
MW: 262.03 g/mol
MDL Number: MFCD12962584
CAS Number: 1190319-92-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR