2-Iodo-1H-pyrrolo[2,3-b]pyridine ≥95%

Supplier: Matrix Scientific

076577-250MG 076577-1G
102546-926EA 967.69 USD
102546-926 102546-928
2-Iodo-1H-pyrrolo[2,3-b]pyridine ≥95%
2-Iodo-1H-pyrrolo[2,3-b]pyridine

Hazard: Irritant

Formula: C₇H₅IN₂
MDL Number: MFCD16875827
CAS Number: 1227270-32-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR