N-[(S)-1-Ethoxycarbonyl-3-phenylpropyl]-L-alanine ≥95%

Supplier: Matrix Scientific

Synonyms: N-[(S)-(+)-1-Ethoxycarbonyl-3-phenylpropyl]-L-alanine, NEPA, N-[(S)-1-Carboxyethyl]-L-homophenylalanine Ethyl Ester, (S)-2-(((S)-1-Ethoxy-1-oxo-4-phenylbutan-2-yl)amino)propanoic acid, N-(1S)-Ethoxycarbonyl-3-phenylpropyl-(S)-alanine

076576-5G 076576-10G 076576-100G
102546-920EA 55.38 USD
102546-920 102546-922 102546-924
N-[(S)-1-Ethoxycarbonyl-3-phenylpropyl]-L-alanine ≥95%
N-[(S)-1-Ethoxycarbonyl-3-phenylpropyl]-L-alanine

Hazard: Irritant

Formula: C₁₅H₂₁NO₄
MW: 279.34 g/mol
MDL Number: MFCD00209976
CAS Number: 82717-96-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR