Methyl-5-bromo-2-methyl-3-nitrobenzoate ≥95%

Supplier: Matrix Scientific

Synonyms: Methyl 5-bromo-2-methyl-3-nitrobenzoate

076573-1G 076573-5G 076573-10G
102546-902EA 233.85 USD
102546-902 102546-904 102546-906
Methyl-5-bromo-2-methyl-3-nitrobenzoate ≥95%
Methyl-5-bromo-2-methyl-3-nitrobenzoate

Hazard: Irritant

Formula: C₉H₈BrNO₄
MW: 274.08 g/mol
MDL Number: MFCD07781280
CAS Number: 220514-28-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR