(S)-Butyl-2-aminopropanoate ≥95%

Supplier: Matrix Scientific

Synonyms: H-Ala-OBu, (S)-Butyl 2-aminopropanoate

076572-1G 076572-5G 076572-25G
102546-896EA 93.85 USD
102546-896 102546-898 102833-926
(S)-Butyl-2-aminopropanoate ≥95%
(S)-Butyl-2-aminopropanoate

Hazard: Irritant

Formula: C₇H₁₅NO₂
MW: 145.2 g/mol
Boiling Pt: 72…43 °C (10 Torr)
MDL Number: MFCD12796122
CAS Number: 2885-02-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR