7-Bromoquinoline-2-carboxylic acid ≥95%

Supplier: Matrix Scientific

076571-250MG 076571-1G
102546-892EA 826.15 USD
102546-892 102546-894
7-Bromoquinoline-2-carboxylic acid ≥95%
7-Bromoquinoline-2-carboxylic acid

Hazard: Irritant

Formula: C₁₀H₆BrNO₂
MDL Number: MFCD11108693
CAS Number: 1057217-63-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR