(S)-Methyl-2-amino-3-((tert-butoxycarbonyl)-amino)propanoate ≥95%
Supplier: Matrix Scientific
Synonyms:
(S)-Methyl 2-amino-3-((tert-butoxycarbonyl)amino)propanoate
076568-250MG
076568-1G
076568-5G
102546-876EA
301
USD
102546-876
102546-878
102546-880
(S)-Methyl-2-amino-3-((tert-butoxycarbonyl)-amino)propanoate ≥95%
(S)-Methyl-2-amino-3-((tert-butoxycarbonyl)-amino)propanoate
Hazard: Irritant/Refrigerate
Formula:
C₉H₁₈N₂O₄ MW: 218.25 g/mol |
MDL Number:
MFCD04039602 CAS Number: 77087-60-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...