4-Methyl-5-nitro-1H-indazole ≥95%

Supplier: Matrix Scientific

076565-250MG 076565-1G
102546-860EA 924.62 USD
102546-860 102546-862
4-Methyl-5-nitro-1H-indazole ≥95%
4-Methyl-5-nitro-1H-indazole

Hazard: Irritant

Formula: C₈H₇N₃O₂
MW: 177.1600 g/mol
MDL Number: MFCD03787902
CAS Number: 101420-67-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR