tert-Butyl-2-(aminomethyl)azetidine-1-carboxylate ≥95%

Supplier: Matrix Scientific

Synonyms: 2-Aminomethyl-azetidine-1-carboxylic acid tert-butyl ester, tert-Butyl 2-(aminomethyl)azetidine-1-carboxylate

076564-250MG 076564-1G
102546-856EA 826.15 USD
102546-856 102546-858
tert-Butyl-2-(aminomethyl)azetidine-1-carboxylate ≥95%
tert-Butyl-2-(aminomethyl)azetidine-1-carboxylate

Hazard: Irritant

Formula: C₉H₁₈N₂O₂
MW: 186.26 g/mol
MDL Number: MFCD08544502
CAS Number: 939760-37-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR