3,5-Dibromo-1-methylpyridin-2(1H)-one ≥95%

Supplier: Matrix Scientific

076563-250MG 076563-1G
102546-852EA 476.92 USD
102546-852 102546-854
3,5-Dibromo-1-methylpyridin-2(1H)-one ≥95%
3,5-Dibromo-1-methylpyridin-2(1H)-one

Hazard: Irritant

Formula: C₆H₅Br₂NO
MDL Number: MFCD00464328
CAS Number: 14529-54-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR