4-(Methylamino)-2-(methylthio)pyrimidine-5-carbaldehyde ≥95%

Supplier: Matrix Scientific

076561-250MG 076561-1G 076561-5G
102546-842EA 427.69 USD
102546-842 102546-844 102546-846
4-(Methylamino)-2-(methylthio)pyrimidine-5-carbaldehyde ≥95%
4-(Methylamino)-2-(methylthio)pyrimidine-5-carbaldehyde

Hazard: Irritant

Formula: C₇H₉N₃OS
MW: 183.23 g/mol
MDL Number: MFCD04115117
CAS Number: 185040-32-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR