Ethyl-3-ethoxy-3-iminopropanoate hydrochloride ≥95%

Supplier: Matrix Scientific

Synonyms: Ethyl 3-ethoxy-3-iminopropionate hydrochloride

076560-100G 076560-25G
102546-840EA 276.92 USD
102546-840 102552-038
Ethyl-3-ethoxy-3-iminopropanoate hydrochloride ≥95%
Ethyl-3-ethoxy-3-iminopropanoate hydrochloride

Hazard: Irritant

Formula: C₇H₁₄ClNO₃
MW: 195.65 g/mol
MDL Number: MFCD00051405
CAS Number: 2318-25-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR