Furan-2,5-diyldimethanol ≥95%

Supplier: Matrix Scientific

Synonyms: 2,5-Furandimethanol, 5-(Hydroxymethyl)furfuryl Alcohol

076559-1G 076559-5G
102546-832EA 80 USD
102546-832 102546-834
Furan-2,5-diyldimethanol ≥95%
Furan-2,5-diyldimethanol

Hazard: Irritant/Refrigerate

Formula: C₆H₈O₃
Boiling Pt: 95…120 °C (0.4 Torr)
Melting Pt: 80 °C
MDL Number: MFCD00003253
CAS Number: 1883-75-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR