1,5-Dichloro-3-methyl-2-nitrobenzene ≥95%

Supplier: Matrix Scientific

076552-250MG 076552-1G 076552-5G
102546-796EA 400 USD
102546-796 102546-798 102546-800
1,5-Dichloro-3-methyl-2-nitrobenzene ≥95%
1,5-Dichloro-3-methyl-2-nitrobenzene

Hazard: Irritant

Formula: C₇H₅Cl₂NO₂
MW: 206.03 g/mol
MDL Number: MFCD11878022
CAS Number: 118665-00-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR