3-Benzoylpyridine ≥95%

Supplier: Matrix Scientific

Synonyms: Phenyl 3-pyridyl ketone, Phenyl(pyridin-3-yl)methanone

076551-5G 076551-10G 076551-100G
102546-790EA 107.69 USD
102546-790 102546-792 102546-794
3-Benzoylpyridine ≥95%
3-Benzoylpyridine

Hazard: Irritant

Formula: C₁₂H₉NO
MW: 183.21 g/mol
MDL Number: MFCD00006394
CAS Number: 5424-19-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR