5,6-Dibromopyridin-3-amine ≥95%

Supplier: Matrix Scientific

Synonyms: 5-Amino-2,3-dibromopyridine

076550-250MG 076550-1G 076550-5G
102546-784EA 498.46 USD
102546-784 102546-786 102546-788
5,6-Dibromopyridin-3-amine ≥95%
5,6-Dibromopyridin-3-amine

Hazard: Irritant

Formula: C₅H₄Br₂N₂
MW: 251.9100 g/mol
MDL Number: MFCD11110698
CAS Number: 53242-19-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR