6-Aminopyridine-2-carbonitrile ≥95%

Supplier: Matrix Scientific

Synonyms: 6-Aminopicolinonitrile

076549-250MG 076549-1G
102546-780EA 301 USD
102546-780 102546-782
6-Aminopyridine-2-carbonitrile ≥95%
6-Aminopyridine-2-carbonitrile

Hazard: Irritant

Formula: C₆H₅N₃
MW: 119.1200 g/mol
MDL Number: MFCD11044252
CAS Number: 370556-44-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR