N-Methylcyclopropanamine hydrochloride ≥95%

Supplier: Matrix Scientific

076546-1G 076546-5G 076546-10G
102546-764EA 340 USD
102546-764 102546-766 102546-768
N-Methylcyclopropanamine hydrochloride ≥95%
N-Methylcyclopropanamine hydrochloride

Hazard: Irritant

Formula: C₄H₁₀ClN
MDL Number: MFCD09997746
CAS Number: 67376-94-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR