Dipyridin-2-ylcarbonate ≥95%
Supplier: Matrix Scientific
076540-5G
076540-25G
102546-740EA
70.77
USD
102546-740
102552-036
Dipyridin-2-ylcarbonate ≥95%
Dipyridin-2-ylcarbonate
Hazard: Irritant
Formula:
C₁₁H₈N₂O₃ Melting Pt: 90 °C |
MDL Number:
MFCD00191407 CAS Number: 1659-31-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...