(S)-2-((tert-Butoxycarbonyl)amino)-2-cyclopentylacetic acid ≥95%

Supplier: Matrix Scientific

Synonyms: Boc-Cpg-OH·DCHA

076539-1G 076539-5G
102546-734EA 640 USD
102546-734 102546-736
(S)-2-((tert-Butoxycarbonyl)amino)-2-cyclopentylacetic acid ≥95%
(S)-2-((tert-Butoxycarbonyl)amino)-2-cyclopentylacetic acid

Hazard: Irritant

Formula: C₁₂H₂₁NO₄
MW: 243.3 g/mol
MDL Number: MFCD05663437
CAS Number: 109183-72-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR