1-tert-Butyl-3-ethyl-4-aminopiperidine-1,3-dicarboxylate ≥95%

Supplier: Matrix Scientific

Synonyms: 1-tert-Butyl 3-ethyl 4-aminopiperidine-1,3-dicarboxylate

076537-250MG 076537-1G
102546-726EA 826.15 USD
102546-726 102546-728
1-tert-Butyl-3-ethyl-4-aminopiperidine-1,3-dicarboxylate ≥95%
1-tert-Butyl-3-ethyl-4-aminopiperidine-1,3-dicarboxylate

Hazard: Irritant

Formula: C₁₃H₂₄N₂O₄
MW: 272.3407 g/mol
MDL Number: MFCD11877860
CAS Number: 932035-01-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR