Methyl-4-hydroxy-3-(trifluoromethyl)benzoate ≥95%

Supplier: Matrix Scientific

Synonyms: Methyl 4-hydroxy-3-(trifluoromethyl)benzoate

076535-1G 076535-5G 076535-10G
102546-716EA 66.15 USD
102546-716 102546-718 102546-720
Methyl-4-hydroxy-3-(trifluoromethyl)benzoate ≥95%
Methyl-4-hydroxy-3-(trifluoromethyl)benzoate

Hazard: Irritant

Formula: C₉H₇F₃O₃
MW: 220.1500 g/mol
MDL Number: MFCD06204091
CAS Number: 115933-50-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR