4-Cbz-aminopiperidine ≥95%

Supplier: Matrix Scientific

Synonyms: 4-Benzyloxycarbonylaminopiperidine, 4-Cbz-Aminopiperidine

076533-1G 076533-5G
102546-706EA 196.92 USD
102546-706 102546-708
4-Cbz-aminopiperidine ≥95%
4-Cbz-aminopiperidine

Hazard: Irritant

Formula: C₁₃H₁₈N₂O₂
MW: 234.2942 g/mol
MDL Number: MFCD04038571
CAS Number: 182223-54-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR