3-Chlorosulfonyl-4-methylbenzoic acid ≥95%

Supplier: Matrix Scientific

076531-1G 076531-5G
102546-698EA 161.54 USD
102546-698 102546-700
3-Chlorosulfonyl-4-methylbenzoic acid ≥95%
3-Chlorosulfonyl-4-methylbenzoic acid

Hazard: Irritant

Formula: C8H7ClO4S
MW: 234.6600 g/mol
MDL Number: MFCD00625710
CAS Number: 2548-29-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR