2,4-Dichloro-3-cyanopyridine ≥95%

Supplier: Matrix Scientific

Synonyms: 2,4-Dichloropyridine-3-carbonitrile, 2,4-Dichloronicotinonitrile

076530-1G 076530-5G
102546-694EA 889.23 USD
102546-694 102546-696
2,4-Dichloro-3-cyanopyridine ≥95%
2,4-Dichloro-3-cyanopyridine

Hazard: Irritant

Formula: C₆H₂Cl₂N₂
MW: 172.996 g/mol
MDL Number: MFCD09750302
CAS Number: 180995-12-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR